missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Multiplex Master Mix

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).

$340.00 - $11,560.00

Specifications

Concentration 2X
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method Multiplex qPCR
Polymerase AmpliTaq Fast DNA Polymerase
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
4461881
Encompass
View Documents Promotion Details
Applied Biosystems™
4461881
1 mL
Each for $340.00
Only null left
 
4461882
Encompass
View Documents Promotion Details
Applied Biosystems™
4461882
5 mL
Each for $1,532.00
Only null left
 
4461884
Encompass
View Documents Promotion Details
Applied Biosystems™
4461884
2 x 5 mL
Each for $2,735.00
Only null left
 
4484262
Encompass
View Documents Promotion Details
Applied Biosystems™
4484262
5 x 5 mL
Each for $6,310.00
Only null left
 
4484263
Encompass
View Documents Promotion Details
Applied Biosystems™
4484263
10 x 5 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4486295
Encompass
View Documents Promotion Details
Applied Biosystems™
4486295
50 mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
  • Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
  • Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility
Specifications

Specifications

2X
High
AmpliTaq Fast DNA Polymerase
TaqMan
Gene Expression, Virus Detection
DNA
Primer-probe
Multiplex qPCR
Fast
Multiplex Master Mix
2 X
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.