missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Multiplex Master Mix

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).

Supplier:  Applied Biosystems™ 4484263

Encompass

 View more versions of this product

Catalog No. 4484263

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
  • Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
  • Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility
Specifications

Chemical Identifiers

2X
High
AmpliTaq Fast DNA Polymerase
TaqMan
Gene Expression, Virus Detection
DNA
Primer-probe
Multiplex qPCR
Fast
Multiplex Master Mix
2 X

Specifications

2X
Primer-probe
Multiplex qPCR
Fast
TaqMan
10 x 5 mL
2 X
DNA
Contains 10 bottles (5 mL each) of 2X TaqMan™ Multiplex Master Mix sufficient for 5000 reactions.


TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer.

Store in refrigerator (2–8°C).

High
AmpliTaq Fast DNA Polymerase
7500 Fast System, 7500 System, QuantStudio™, ViiA™ 7 System
Multiplex Master Mix
Gene Expression, Virus Detection
5000 Reactions
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.