Learn More
Applied Biosystems™ High-Capacity cDNA Reverse Transcription Kit
The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction.
$732.00 - $5,400.00
Specifications
| Concentration | 50 U/μL |
|---|---|
| Format | Kit |
| Reaction Speed | 120 min. |
| Technique | Reverse Transcription |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Includes | No. of Reactions | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | No. of Reactions | Price | Quantity & Availability | ||||
4368814
|
Applied Biosystems™
4368814 |
Kit only | 200 Reactions |
Each for $732.00
|
|
||||
4374966
|
Applied Biosystems™
4374966 |
Kit with Rnase Inhibitor | 200 Reactions |
Each for $1,264.00
|
|
||||
4368813
|
Applied Biosystems™
4368813 |
Kit only | 1,000 Reactions |
Each for $2,945.00
|
|
||||
4374967
|
Applied Biosystems™
4374967 |
Kit with Rnase Inhibitor | 1,000 Reactions |
Each for $5,400.00
|
|
||||
Description
The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction. Both types of kit deliver extremely high-quality, single-stranded cDNA from 0.02–2 μg total RNA. Reactions can be scaled up to 100 μL to generate 10 μg of cDNA from a single reaction.
The High Capacity cDNA Reverse Transcription Kit with RNase Inhibitor includes the recombinant enzyme used to prevent degradation of RNA templates and lacks DNA endonuclease activity for better product yield.
In addition to their use in RT-PCR, this kit may be used in traditional PCR, microarrays, and for the generation of cDNA archives.
Features of the High Capacity cDNA Reverse Transcription Kit include:
- Linear target amplification for real-time PCR
- Higher yields and precision than other cDNA synthesis kits at a fraction of the cost
- 10-fold greater dynamic range than other kits
Extensively tested with a variety of templates
Quantitative first-strand synthesis of all RNA species is achieved with the use of the random primers. This kit has been extensively tested with a variety of RNA templates, including GC- and AU-rich targets and RNAs expressed at low levels. cRNA can also be efficiently generated from in vitro transcription of cDNA.
Order Info
Shipping Condition: Dry ice
Specifications
| 50 U/μL | |
| 120 min. | |
| 1 Ea. | |
| Yes | |
| First-Strand cDNA | |
| Reverse Transcription |
| Kit | |
| Reverse Transcription | |
| MultiScribe | |
| Dry Ice | |
| Separate components | |
| RNA |
For Research Use Only. Not for use in diagnostic procedures.