missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ High-Capacity cDNA Reverse Transcription Kit

Catalog No. 4368813
Encompass
Change view
Click to view available options
Includes:
Kit only
Kit with Rnase Inhibitor
No. of Reactions:
1,000 Reactions
200 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes No. of Reactions
4368813 Kit only 1,000 Reactions
4368814 Kit only 200 Reactions
4374966 Kit with Rnase Inhibitor 200 Reactions
4374967 Kit with Rnase Inhibitor 1,000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 4368813 Supplier Applied Biosystems™ Supplier No. 4368813

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction.

The High Capacity cDNA Reverse Transcription Kit, available with or without RNase Inhibitor, contains all components necessary for the quantitative conversion of up to 2 μg of total RNA into single-stranded cDNA in a single 20 μL reaction. Both types of kit deliver extremely high-quality, single-stranded cDNA from 0.02–2 μg total RNA. Reactions can be scaled up to 100 μL to generate 10 μg of cDNA from a single reaction.

The High Capacity cDNA Reverse Transcription Kit with RNase Inhibitor includes the recombinant enzyme used to prevent degradation of RNA templates and lacks DNA endonuclease activity for better product yield.

In addition to their use in RT-PCR, this kit may be used in traditional PCR, microarrays, and for the generation of cDNA archives.

Features of the High Capacity cDNA Reverse Transcription Kit include:

  • Linear target amplification for real-time PCR
  • Higher yields and precision than other cDNA synthesis kits at a fraction of the cost
  • 10-fold greater dynamic range than other kits

Extensively tested with a variety of templates

Quantitative first-strand synthesis of all RNA species is achieved with the use of the random primers. This kit has been extensively tested with a variety of RNA templates, including GC- and AU-rich targets and RNAs expressed at low levels. cRNA can also be efficiently generated from in vitro transcription of cDNA.

Order Info

Shipping Condition: Dry ice

Specifications

Concentration 50 U/μL
Content And Storage

• 10X RT Buffer (2 x 1 mL)
• 10X RT Random Primers (2 x 1 mL)
• 25X dNTP Mix (1.0 mL at 100 mM)
• MultiScribe Reverse Transcriptase (1.0 mL at 50 U/μL)

Store at –15 to –25°C.
 

Format Kit
Reaction Speed 120 min.
Technique Reverse Transcription
Quantity 1 Ea.
Reverse Transcriptase MultiScribe
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 1,000 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) 7 kb
Starting Material RNA
Includes Kit only
Show More Show Less
Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?

We recommend the following kits:

  • High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
  • High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).

What is the concentration of the random primers in the High-Capacity cDNA Reverse Transcription Kit (Cat. no. 4368813)?

The concentration of the "random primers" in the High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4368813) is 25 µM. We recommend that you use these primers at a 1X concentration (2.5 µM) in reverse transcription reactions.

Is the 25X 100 mM dNTP Mix included in the High Capacity cDNA Reverse Transcription Kit (Cat. No. 4368813, 4368814) 100 mM each or 100 mM combined?

It contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).

For Research Use Only. Not for use in diagnostic procedures.