missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ 4 BpiI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
$240.00 - $962.00
Spécifications
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
|---|---|
| Methylation Sensitivity | Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive |
| Enzyme | Bpi I |
| Compatible Buffer | Anza™ 10X Red Buffer, Anza™ 10X Buffer |
| Sensitive to Heat Inactivation | Yes |
Comprend: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
IVGN0044
|
Invitrogen™
IVGN0044 |
250 units |
chaque for $240.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
IVGN0046
|
Invitrogen™
IVGN0046 |
1200 units |
chaque for $962.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Spécifications
| Approved for shipment on Wet or Dry Ice | |
| Bpi I | |
| Yes | |
| Restriction Enzyme Cloning System | |
| Anza |
| Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
| Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
| 37°C | |
| Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.