missing translation for 'onlineSavingsMsg'
Learn More

Antimony ICP-MS Standard, 1000 ppm Sb in H2O/Tartaric Acid/tr HNO3, Ricca Chemical

Catalog No. MSSB1KW100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSSB1KW100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSSB1KW100 Supplier Ricca Chemical Company Supplier No. MSSB1KW100

Chemical Identifiers

CAS 7440-36-0 , 7697-37-2 , 7647-01-0 , 87-69-4 , 7732-18-5
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sb
CAS Min % 96.0, 0.72, 0.57, 0.14, 0.1
CAS Max % 100.0, 0.75, 0.59, 0.15, 0.1
Equivalence 1 mL = 1 mg Sb
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Antimony ICP-MS Standard
Show More Show Less