missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antifoam A, Spectrum™ Chemical
A1302
Chemical Identifiers
| CAS | 63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6 |
|---|---|
| Molecular Formula | (C2H6OSi)n |
| Molecular Weight (g/mol) | NaN |
| MDL Number | MFCD00132673 |
| IUPAC Name | Polydimethylsiloxane |
| SMILES | C[Si](C)(-*)O-* |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18600723
|
N/A | 100 mL | Poly Bottle |
N/A
|
|
||||
|
18600724
|
N/A | 500 mL | Poly Bottle |
N/A
|
|
||||
|
18600725
|
N/A | 4 L | Poly Bottle |
N/A
|
|
||||
|
18600727
|
N/A | 20 L | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18600726
|
N/A | 200 L | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Chemical Identifiers
| 63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6 | |
| NaN | |
| Polydimethylsiloxane |
| (C2H6OSi)n | |
| MFCD00132673 | |
| C[Si](C)(-*)O-* |
Specifications
| 63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6 | |
| (C2H6OSi)n | |
| C[Si](C)(-*)O-* | |
| NaN | |
| Antifoam A |
| 80% to 100%, 10% to 20%, 5% to <10%, 3% to <5%, 1% to <3% | |
| MFCD00132673 | |
| Polydimethylsiloxane | |
| Ungraded |