Learn More
Antifoam A, Spectrum™ Chemical
A1302
Supplier: Spectrum Chemical Mfg Cor A13024LTPL
| Quantity | 4 L |
|---|---|
| Packaging | Poly Bottle |
Description
Spectrum™ Chemical Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Chemical Identifiers
| 63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6 | |
| NaN | |
| Polydimethylsiloxane |
| (C2H6OSi)n | |
| MFCD00132673 | |
| C[Si](C)(-*)O-* |
Specifications
| Antifoam A | |
| 80% to 100%, 10% to 20%, 5% to <10 %, 3% to <5%, 1% to <3 % | |
| Poly Bottle | |
| MFCD00132673 | |
| Polydimethylsiloxane | |
| Ungraded |
| 63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6 | |
| 4 L | |
| (C2H6OSi)n | |
| C[Si](C)(-*)O-* | |
| NaN |