missing translation for 'onlineSavingsMsg'
Learn More

Antifoam A, Spectrum™ Chemical

A1302

Supplier:  Spectrum Chemical Mfg Cor A1302500MLPL

Quantity 500 mL
Packaging Poly Bottle

 View more versions of this product

Catalog No. 18600724


Only null left

Description

Description

Spectrum™ Chemical Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Chemical Identifiers

63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6
NaN
Polydimethylsiloxane
(C2H6OSi)n
MFCD00132673
C[Si](C)(-*)O-*

Specifications

Antifoam A
80% to 100%, 10% to 20%, 5% to <10%, 3% to <5%, 1% to <3%
Poly Bottle
MFCD00132673
Polydimethylsiloxane
Ungraded
63148-62-9 , 67762-90-7 , 556-67-2 , 541-02-6 , 69430-40-6
500 mL
(C2H6OSi)n
C[Si](C)(-*)O-*
NaN
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions