missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 23-193 Heavy Duty Foam-Insulated PVC-Coated Gloves

Catalog No. 2705565
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
2705565 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2705565 Supplier Ansell Supplier No. 2319311100

missing translation for 'validMainframeMsg'

PVC coating remains flexible, and helps users maintain a secure grip in even the lowest temperatures.

  • PVC vinyl coating protects hands from abrasions, while also remaining soft and flexible during cold applications
  • Coating repels water, oil, and grease
  • Foam insulation helps keep hands warm in low temperatures
  • Enhanced grip for safe handling of heavy duty objects
  • Curved fingers, wing thumb, and soft jersey cotton lining

Specifications

ANSI Abrasion Rating ANSI 3
Color Orange
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material PVC
Size 10
EN388 Standard 3121B
For Use With (Application) Metal Fabrication
Glove Cuff Style Safety
Liner Material Cotton Jersey
Series 23-193
Show More Show Less