missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 92-605 Disposable Chemical Resistant Nitrile Gloves
GREENER_CHOICE

Catalog No. 19148541
Change view
Click to view available options
Size:
6.5/7
7.5/8
8.5/9
9.5/10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19148541 7.5/8
19148540 6.5/7
19148542 8.5/9
19148543 9.5/10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19148541 Supplier Ansell Supplier No. 9260500M
Edge

Disposable, nitrile gloves with extended cuff features splash protection against hazardous chemicals.

  • Does not contain natural rubber latex proteins
  • Strong grip in wet or dry conditions
  • Rolled beaded for easy donning
  • Powder free, Polymer-coated
  • Ambidextrous
  • Textured Fingers
  • AQL: 1.5

Specifications

Chlorinated Yes
AQL 1.5
Color Green
Disposable Single-use
Finger Thickness (English) 6.2 mil
Finger Thickness (Metric) 0.16 mm
Latex Free Yes
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Size 7.5/8
EN 421 Yes
EN ISO 374-1 Type B
Glove Cuff Style Beaded/Rolled
Show More Show Less