missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves

Catalog No. 19823455
Change view
Click to view available options
Size:
10
11
6
8
9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19823455 11
19823450 6
19823452 8
19823453 9
19823454 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19823455 Supplier Ansell Supplier No. 8081311110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flame resistant gloves provide Arc Flash Hazard/Risk Category 2 protection against electrical arc flash hazards. Aramid Fiber shell allows for safe handling of sharp objects.

  • Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
  • Neoprene foam coating enhances grip in dry, wet, or oily conditions
  • Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
  • Ergonomic design is comfortable and flexible

Specifications

ANSI Abrasion Rating ANSI 3
Arc Flash Level 2
Certifications/Compliance NFPA 70E Hazard Risk Category 2
Certifications/Compliance (Europe) EN 407: 412110
Construction Knit
Coating Material Neoprene
Color Green, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material Aramid Fiber
Product Type Arc Flash Gloves
Size 11
EN388 Standard 2X42C
For Use With (Application) Automotive Assembly, Equipment/Machinery Operation, Mining, and Utility Work
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series ActivArmr™ 80-813
Thumb Style Wing
Yarn Gauge 13
Length (English) 8.86 to 10.24 in.
Show More Show Less