Learn More
Ansell ActivArmr™ 47-402 Nitrile Coated Medium Duty Gloves
Nitrile coating is abrasion and oil-resistant to enhance grip capabilities in dry or oily conditions. Gloves feature interlock knit lining, generous sizing, and a contoured design for increased comfort.
Supplier:  Ansell 4740211100
 View more versions of this product
Catalog No. 19-019880
Only null left
Explore available promotions
Description
Description
  • ActivArmr™ gloves combine the comfort of cotton gloves with the abrasion resistance and oil repellence of a nitrile-coated glove
  • Interlock knit lining, generous sizing, and contoured designs increase comfort
  • Special nitrile formulation provides mechanical resistance and oil repellency
  • Designed to conform to the hand's natural shape for long wear periods
  • Ensure cleanliness of metal sheets in the steel and motor industries
  • Ideal for a variety of medium-duty job tasks
Specifications
Specifications
ANSI 4
Cut and Sewn
Blue, Natural
Specific
Cotton
10
Knit
Polybag
47-402
Full Glove
Nitrile
No Texture
Yes
Dip-coated Gloves
Automotive Repairs, Heavy Equipment Operation, Metal Fabrication, and Chemical Handling
Smooth
ANSI/ISEA 105-16 Level 2
Wing
Product Suggestions
Product Suggestions