Learn More
Ansell HyFlex™ 11-920 Nitrile Coated Oil Repellent Gloves
Seamless knit gloves feature ANSELL GRIP™ Technology which utilizes microscopic channels in the glove coating to displace oil, enhancing grip capabilities when working with oily parts.
Supplier: Ansell 1192011090
Description
- High performance grip and oil repellence when working in oily environments
- ANSELL GRIP™ Technology contains microscopic channels in the glove coating to displace oil
- RIPEL™ Liquid Repellence Technology creates a liquid impermeable layer to prevent oil or lubricant from making contact with the wearer's skin
- Seamless knit design provides a comfortable fit with maximum dexterity
Specifications
| ANSI 4 | |
| ANSI A1 | |
| Specific | |
| Large | |
| Knit | |
| 15 |
| Seamless Knit | |
| No Texture | |
| Nylon | |
| Automotive Assembly, Equipment Operation, Machinery, and Metal Fabrication | |
| 11-920 |