missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin

Catalog No. AAJ6097714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6097714 25 g
AAJ6097706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6097714 Supplier Thermo Scientific Chemicals Supplier No. J6097714

Ampicillin, CAS # 69-53-4, is structurally related to penicillin and is considered to be a semi-synthetic antibiotic with antibacterial activity. | CAS: 69-53-4 | C16H19N3O4S | 349.405 g/mol

Ampicillin is used as asemi-synthetic antibiotic which has antibacterial activity structurally related to penicillin. It is widely used selection reagent for transformed cells expressing β-lactamase. It is also used to prevent and treat a number of bacterial infections. It may also be used to prevent group B streptococcal infection in newborns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ampicillin is a broad-spectrum, semi-synthetic, β-lactam penicillin with bactericidal activity. It acts against several gram-positive and -negative infections
  • This compound binds to and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall. This inactivation interferes with the cross-linkage of peptidoglycan chains essential for bacterial cell wall strength and rigidity. Consequently, there is an interruption in the bacterial cell wall synthesis which results in the weakening of the bacterial cell wall causing cell lysis

Application

  • In laboratory experiments, it is commonly used as a selector reagent for transformed cells expressing β-lactamase. The ampicillin-resistant gene (ampR) catalyzes the hydrolysis of the β-lactam ring of ampicillin and naturally detoxifies the drug
  • Ampicillin is commonly used as a selection marker for E. coli and other bacteria during plasmid DNA isolation, protein expression, and gene cloning

Chemical Identifiers

CAS 69-53-4
Molecular Formula C16H19N3O4S
Molecular Weight (g/mol) 349.405
MDL Number MFCD00005175
InChI Key AVKUERGKIZMTKX-NJBDSQKTSA-N
PubChem CID 6249
ChEBI CHEBI:28971
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C

Specifications

Type Ampicillin
Physical Form Powder
Melting Point 208°C (decomposition)
Color White
Quantity 25 g
Beilstein 1090925
Merck Index 14,586
Solubility Information In 1 M NH4OH,50mg/ml
Formula Weight 349.4
Chemical Name or Material Ampicillin
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 200-709-7
RTECSNumber XH8350000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only