missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin sodium salt

Catalog No. AAJ6380709
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6380709 10 g
AAJ6380706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6380709 Supplier Thermo Scientific Chemicals Supplier No. J6380709

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ampicillin sodium salt, CAS # 69-52-3, is the sodium salt form of ampicillin, a semisynthetic derivative of aminopenicillin compound that shows antibacterial activity against both gram-positive and gram-negative bacteria. | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol

Ampicillin sodium salt is a reagent for transformed cells expressing beta-lactamase. It acts as a structural analogue of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme and prevents the cross-linking of the peptidoglycan of the cell wall necessary for the growth of the bacterium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Ampicillin sodium salt is a semi-synthetic derivative of penicillin with a broad-spectrum antibacterial activity against several species, including Bacillus subtilis, Staphylococcus aureus, Escherichia coli, and Ralstonia solanacearum

• It can inhibit bacterial cell wall synthesis by binding to penicillin-binding proteins, inhibiting peptidoglycan synthesis, which is an essential component of the bacterial cell wall

Application

• Ampicillin sodium salt has been used as a reagent for transformed cells that express β-lactamase

• It can be used to select for ampicillin resistance in mutated and transformed cells

• Ampicillin sodium salt acts as a structural analog of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme preventing the cross-linking of the cell wall peptidoglycan, which is necessary for the growth of the bacterium

Chemical Identifiers

CAS 69-52-3
Molecular Formula C16H21N3NaO4S
Molecular Weight (g/mol) 374.411
MDL Number MFCD00064313
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
PubChem CID 131673879
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]

Specifications

Type Ampicillin Sodium Salt
Physical Form Powder
Melting Point 215°C
Color White
Quantity 10 g
Beilstein 4119211
Merck Index 14,586
Solubility Information Freely soluble in water; Sparingly soluble in acetone
Formula Weight 371.39
Chemical Name or Material Ampicillin sodium salt
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P260-P271-P272-P280g-P285-P302+P352-P304+P340-P314-P333+P313-P342+P311-P363-P501c
EINECSNumber 200-708-1
RTECSNumber XH8400000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only