missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iodide, ACS, 99%, Spectrum™ Chemical
Spécifications
Spécifications
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Percent Purity | 99% |
| Grade | ACS |
| Packaging | Amber Glass Bottle |
| Residue after Ignition | 0.0005 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.