missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium acetate, 97+%, MP Biomedicals™
Spécifications
Spécifications
| Color | White |
| Physical Form | Powder or Liquid |
| Quantity | 100 g |
| Assay Percent Range | 0.97+ |
| Formula Weight | 77.08 |
| Density | 1.17 (water = 1) |
| Chemical Name or Material | Ammonium acetate |