missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

$406.87 - $872.29

Specifications

Length (Metric) 4 m
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
View More Specs

Products 9
Catalog Number Mfr. No. Width (Metric) Pore Size Price Quantity  
Catalog Number Mfr. No. Width (Metric) Pore Size Price Quantity  
45-004-000
View Documents
Cytiva
10600000
30 cm 0.1 μm
Pack for $845.28
Only null left
 
45-004-002
View Documents
Cytiva
10600002
30 cm 0.45 μm
Pack for $707.56
Only null left
 
45-004-001
View Documents
Cytiva
10600001
30 cm 0.2 μm
Pack for $872.29
Only null left
 
45-004-005
View Documents
Cytiva
10600005
20 cm 0.1 μm
Pack for $535.67
Only null left
 
45-004-006
View Documents
Cytiva
10600006
20 cm 0.2 μm
Pack for $478.99
Only null left
 
45-004-010
View Documents
Cytiva
10600010
15 cm 0.1 μm
Pack for $473.23
Only null left
 
45-004-012
View Documents
Cytiva
10600012
15 cm 0.45 μm
Pack for $406.87
Only null left
 
45-004-007
View Documents
Cytiva
10600007
20 cm 0.45 μm
Pack for $478.99
Only null left
 
45-004-011
View Documents
Cytiva
10600011
15 cm 0.2 μm
Pack for $406.87
Only null left
 
Description

Description

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Packed in rolls.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

Specifications

Specifications

4 m
Western blots, dot-blot assays, and other protein or nucleic acid methods
Roll
100% Pure Nitrocellulose
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'