Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Supplier:  Cytiva 10600010
 View more versions of this product
Catalog No. 45-004-010
Only null left
Explore available promotions
Description
Description
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Packed in rolls.
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
4 m
Western blots, dot-blot assays, and other protein or nucleic acid methods
100% Pure Nitrocellulose
15 cm
Roll
0.1 μm
Product Suggestions
Product Suggestions