missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls
Description
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Packed in rolls.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
| Length (Metric) | 4 m |
| Width (Metric) | 30 cm |
| For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
| Format | Roll |
| Material | 100% Pure Nitrocellulose |
| Pore Size | 0.45 μm |
| Dimensions (LxW) | 4 m x 30 cm |
| Product Line | Amersham Protran |