missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls

Catalog No. 45004001
Change view
Click to view available options
Width (Metric):
15 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric) Pore Size
45004001 30 cm 0.2 μm
45004000 30 cm 0.1 μm
45004002 30 cm 0.45 μm
45004005 20 cm 0.1 μm
45004006 20 cm 0.2 μm
45004010 15 cm 0.1 μm
45004012 15 cm 0.45 μm
45004007 20 cm 0.45 μm
45004011 15 cm 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 45004001 Supplier Cytiva Supplier No. 10600001
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Packed in rolls.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

Specifications

Length (Metric) 4 m
Width (Metric) 30 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.2 μm
Dimensions (LxW) 4 m x 30 cm
Product Line Amersham Protran