missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Buffer-Saturated Phenol

UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids. The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4.

$242.00 - $748.00

Spécifications

Chemical Name or Material Phenols
Content And Storage Store in refrigerator (2–8°C).
Packaging Type Bottle
pH 7.4
Product Line UltraPure
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
15513039
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
15513039
100 mL
chaque for $242.00
Il en reste null
 
15513047
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
15513047
400mL
chaque for $748.00
Il en reste null
 
Description

Description

  • UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids
  • The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4
  • When mixtures are extracted with UltraPure™ Buffer- Saturated Phenol, proteins are denatured and collect in the organic phase or at the interphase, while most nucleic acids remain in the aqueous phase
  • UltraPure™ Buffer-Saturated Phenol contains no preservatives
  • It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
  • The appearance of the solution is evaluated at room temperature
  • No RNase or DNase activity detected
  • Store in refrigerator at 2°C to 8°C

Order Info

Shipping Condition: Room temperature

Spécifications

Spécifications

Phenols
Bottle
UltraPure
Room Temperature
Store in refrigerator (2–8°C).
7.4
Molecular Biology Grade
Liquid
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.