Learn More
Invitrogen™ SSC (20X), RNase-free
Molecular biology grade, Invitrogen 20X SSC solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use.
Supplier: Invitrogen™ AM9763
Description
Molecular biology grade, Invitrogen 20X SSC solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen™'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Identifiants chimiques
| Store at room temperature. | |
| Bottle | |
| Room Temperature |
| 20X | |
| Ambion | |
| Liquid |
Spécifications
| Store at room temperature. | |
| Bottle | |
| 1L | |
| Liquid |
| 20X | |
| Ambion | |
| Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.