missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FirstChoice™ RLM-RACE Kit

Numéro de catalogue. p-4924873
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 Kit with manual
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AM1700M 1 Kit with manual
AM1700 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AM1700M Fournisseur Invitrogen™ Code fournisseur AM1700M
Il en reste null

Comprend

Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate GAPDH-M, and 0.5M EDTA should be stored at –20°C. Nuclease-free water may be stored at any temperature.

Exige

SuperTaq Thermostable Taq DNA Polymerase sold separately. For optimal amplification of fragments ≥1 kb, use SuperTaq-Plus.

Designed to amplify cDNA only from full-length, capped mRNA, usually producing a single band after PCR

FastAP™ Thermosensitive Alkaline Phosphatase, 10X FastAP Buffer, Tobacco Acid Pyrophosphatase, 10X TAP Buffer, T4 RNA Ligase, 10X T4 RNA Ligase Buffer, M-MLV Reverse Transcriptase, RNase Inhibitor, 10X RT Buffer, 2.5 mM dNTP Mix, Mouse Thymus RNA (1 mg/ml), Random Decamers, 5' RACE Adapter, 3' RACE Adapter, 5' RACE Outer Primer, 5' RACE Inner Primer, 3' RACE Outer Primer, 3' RACE Inner Primer, 5' RACE Outer Control Primer, 5' RACE Inner Control Primer, 5' PCR Control Primer, 3' RACE Control Primer and Ammonium Acetate Stop Solution should be stored at -20°C. Nuclease-free Water may be stored at any temperature.

The FirstChoice™ RLM-RACE Kit is designed to amplify cDNA from full-length, capped mRNA through RNA Ligase Mediated Rapid Amplification of cDNA Ends (RLM-RACE). The kit ensures the amplification of only full-length transcripts by eliminating ribosomal RNA, fragmented mRNA, tRNA, and contaminating genomic DNA during the amplification process. This kit is a major improvement over the basic rapid amplification of cDNA ends (RACE) protocol.

Note: calf intestinal phosphatase (CIP) had been replaced with FastAP Thermosensitive Alkaline Phosphatase adjusted protocol eliminate the need to use phenol/chloroform extraction and ethanol precipitation. If chosen, phenol/chloroform step can be used with updated kit after verifying reaction volumes.

Features of the FirstChoice RLM-RACE Kit include:

  • Selects 5' and/or 3' ends of full-length, capped mRNA
  • Optimized to ensure detection of even the rare mRNA, yielding specific product
  • Allows to finish experiment in less than a day

Selects full-length mRNA—no rRNA, tRNA or degraded mRNA

In the FirstChoice RLM-RACE procedure, full-length mRNAs are selected by treating total or poly(A) RNA with FastAP Thermosensitive Alkaline Phosphatase to remove the 5'-phosphate from all molecules which contain free 5'-phosphates (ribosomal RNA, fragmented mRNA, tRNA, and contaminating genomic DNA). Full-length mRNAs are unaffected. The RNA is then treated with tobacco acid pyrophosphatase (TAP) to remove the cap structure from the full-length mRNA leaving a 5'-monophosphate. A synthetic RNA adapter is ligated to the RNA population—only molecules containing a 5'-phosphate, the uncapped, full-length mRNAs, will accept the adapter. Random-primed, reverse transcription reactions and nested PCR are then performed to amplify the 5'-end of your specific transcript.

From RNA to PCR in less than a day

Each step of the FirstChoice RLM-RACE procedure has been optimized, so you can complete all the enzymatic reactions and even start PCR in no more than 5 hours. The FirstChoice RLM-RACE Kit contains reagents and enzymes to produce 6 RLM-RACE-ready cDNA preparations, primers and nested RACE adapter primers for 100 PCR reactions. Kit includes reverse transcription reagents and 3' RACE Adapter and Primers for 3' RACE. Each kit contains control RNA and primers to test the kit's performance, protocol described in User Guide utilizing Platinum™ II Hot-Start PCR Master Mix.

Order Info

Shipping Condition: Dry ice

Spécifications

Product Type RLM-RACE Kit
Includes Kit with manual, FastAP™ Thermosensitive Alkaline Phosphatase, 10X FastAP Buffer, Tobacco Acid Pyrophosphatase, 10X TAP Buffer, T4 RNA Ligase, 10X T4 RNA Ligase Buffer, M-MLV Reverse Transcriptase, RNase Inhibitor, 10X RT Buffer, 2.5 mM dNTP Mix, Mouse Thymus RNA (1 mg/ml), Random Decamers, 5’ RACE Adapter, 3’ RACE Adapter, 5’ RACE Outer Primer, 5’ RACE Inner Primer, 3’ RACE Outer Primer, 3’ RACE Inner Primer, 5’ RACE Outer Control Primer, 5’ RACE Inner Control Primer, 5’ PCR Control Primer, 3’ RACE Control Primer and Ammonium Acetate Stop Solution, Nuclease-free Water
Content And Storage

•FastAP™ Thermosensitive Alkaline Phosphatase
•10X FastAP Buffer, Tobacco Acid Pyrophosphatase
•10X TAP Buffer, T4 RNA Ligase
•10X T4 RNA Ligase Buffer
•M-MLV Reverse Transcriptase
•RNase Inhibitor
•10X RT Buffer
•2.5 mM dNTP Mix
•Mouse Thymus RNA (1 mg/ml)
•Random Decamers
•5' RACE Adapter
•3' RACE Adapter
•5' RACE Outer Primer
•5' RACE Inner Primer
•3' RACE Outer Primer
•3' RACE Inner Primer
•5' RACE Outer Control Primer
•5' RACE Inner Control Primer
•5' PCR Control Primer
•3' RACE Control Primer
•Ammonium Acetate Stop Solution
•Components should be stored at -20°C. Nuclease-free Water may be stored at any temperature.

Final Product Type cDNA
Optimal Reaction Temperature 42°C
Reverse Transcriptase M-MLV
Format Kit
Product Line Ambion, FirstChoice
For Use With (Application) cDNA Libraries & Library Construction
Quantity 1 Kit with manual

For Research Use Only. Not for use in diagnostic procedures.