Learn More
Invitrogen™ Ammonium Acetate (5 M), RNase-free
Invitrogen Molecular biology grade, 5 M Ammonium Acetate solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use.
Spécifications
| Chemical Name or Material | Ammonium Acetate |
|---|---|
| Content And Storage | Store at room temperature. |
| Concentration | 5 M |
| Packaging Type | Bottle |
| Product Line | Ambion |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
AM9070G
|
Invitrogen™
AM9070G |
100 mL |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
AM9071
|
Invitrogen™
AM9071 |
500 mL |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Invitrogen Molecular biology grade, 5 M Ammonium Acetate solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Spécifications
| Ammonium Acetate | |
| 5 M | |
| Ambion | |
| Room Temperature |
| Store at room temperature. | |
| Bottle | |
| Molecular Biology Grade | |
| Liquid |
For Research Use Only. Not for use in diagnostic procedures.