missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acid-Phenol:Chloroform, pH 4.5 (with IAA, 125:24:1)

In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing.

$184.00 - $404.00

Specifications

Chemical Name or Material Phenols
Content And Storage Store at 4°C or -20°C.
Packaging Type Bottle
pH 4.5
Product Line Ambion
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AM9722
Encompass
View Documents Promotion Details
Invitrogen™
AM9722
400 mL
Each for $404.00
Only null left
 
AM9720
Encompass
View Documents Promotion Details
Invitrogen™
AM9720
100 mL
Each for $184.00
Only null left
 
Description

Description

Acid Phenol:Chloroform:IAA (125:24:1) is premixed and supplied at pH 4.5 ± 0.2. Provided in one bottle of 400 mL. In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality-tested, saturated phenols are ready to use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Phenols
Bottle
Ambion
Room Temperature
Store at 4°C or -20°C.
4.5
Molecular Biology Grade
Liquid
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.