Learn More
Invitrogen™ Acide-Phénol : Chloroforme, pH 4,5 (avec IAA, 125:24:1)
Description
Acid Phenol:Chloroform:IAA (125:24:1) is premixed and supplied at pH 4.5 ± 0.2. Provided in one bottle of 400 mL. In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality-tested, saturated phenols are ready to use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
Order Info
Conditions d’expédition : Température ambiante
Spécifications
Spécifications
| Nom ou substance chimique | Phénols |
| Contenu et stockage | Conserver à 4 °C ou -20 °C. |
| Type d’emballage | Flacon |
| pH | 4.5 |
| Gamme de produit | Ambion |
| Pureté | Grade de biologie moléculaire |
| Quantité | 400 mL |
| Shipping condition (Condition d'expédition) | Température ambiante |
| Forme | Liquide |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.