missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acid-Phenol:Chloroform, pH 4.5 (with IAA, 125:24:1)

In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing.

Supplier:  Invitrogen™ AM9720

Encompass

 View more versions of this product

Catalog No. AM9720


Only null left

Description

Description

Acid Phenol:Chloroform:IAA (125:24:1) is premixed and supplied at pH 4.5 ± 0.2. Provided in one bottle of 400 mL. In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality-tested, saturated phenols are ready to use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Identifiers

Store at 4°C or -20°C.
4.5
Molecular Biology Grade
Liquid
Bottle
Ambion
Room Temperature
Phenols

Specifications

Phenols
Bottle
Ambion
100 mL
Liquid
Store at 4°C or -20°C.
4.5
Molecular Biology Grade
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.