missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hydroxide, extra pure, powder

Numéro de catalogue. AC219130250
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC219130250 25 g Plastic bottle
AC219135000 500 g Plastic bottle
AC219130020 2 kg Plastic bottle
AC219130100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC219130250 Fournisseur Thermo Scientific Chemicals Code fournisseur 219130250
Il en reste null

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Spécifications

Color White
pH 8.5 to 10 (5% aq. suspension)
Physical Form Powder
Quantity 25 g
Linear Formula Al(OH)3
Merck Index 15,338
Solubility Information Solubility in water: insoluble
Formula Weight 78
Percent Purity ≥63.5% (Al2O3)
Grade Extra Pure
Loss on Ignition 36.5% max.
Packaging Plastic bottle
Chemical Name or Material Aluminum hydroxide
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 244-492-7

RUO – Research Use Only