missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 87-320 Rubber Gloves

Numéro de catalogue. 17609979
Change view
Click to view available options
Size:
10
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size
17609979 10
17609976 7
17609977 8
17609978 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 17609979 Fournisseur Ansell Code fournisseur 87320100*
Il en reste null

Flocked-lined natural rubber gloves, combining comfort, flexibility and food compliance.

Key Features and Benefits

  • Beaded cuff: For easy-to-wear chemical gloves with tear resistance
  • Food-safe: Excellent hand and food safety gloves
  • Good abrasion resistance: Protective gloves for use in low-hazard areas

Description

  • Increased protection and convenience: Flock-lined and lightweight, AlphaTec™ 87-320 rubber work gloves come with beaded cuffs, which ensure tear resistance and easy donning and doffing
  • Food handling compliance: Featuring an AQL of 1.5, these orange food handling gloves are suitable for a wide range of food applications
  • Enhanced durability: AlphaTec™ 87-320 hand protection gloves are also abrasion-resistant, lending themselves to low-hazard tasks
  • Increased comfort: Owing to their 100% cotton flock lining, these flexible work gloves are comfortable and offer wearers the required flexibility for light-duty tasks

Spécifications

Certifications/Compliance EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact, Category III
Certifications/Compliance (Europe) CE 0493, EN 388:2016 +A1:2018, REACH Compliant
Chlorinated No
Applications Industrial
Color Orange
Finger Thickness (English) 18 mil
Finger Thickness (Metric) 0.45 mm
Latex Free No
Material Rubber Latex
Palm Thickness (English) 18 mil
Palm Thickness (Metric) 0.45 mm
Product Type Chemical Protection Gloves
Size 10
Thickness (English) 18 mil
Thickness (Metric) 0.45 mm
EN388 Standard EN 388:2016 +A1:2018 - 2010X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Beaded
Grip Design Raised Diamond
Liner Material Cotton Flock
Lining Flock Lined
Length (English) 11.8 in.
Length (Metric) 300 mm
Afficher plus de résultats Afficher moins de résultats