Learn More
AlphaProTech Critical Cover™ SafeStep™ Shoe Covers
Catalog No. 19125445
Change view
Click to view available options
Size:
Universal
Medium
Large
X-Large
Thread Color:
Black
Orange
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Color
19125445 X-Large Black
19125442 Medium Yellow
19125443 Large Orange
19125444 Universal White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19125445 Supplier AlphaProTech Supplier No. SH E1713 BH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High-performing, environmentally friendly shoe cover features inherent non-skid physical properties. Choose from four sizes to ensure a proper fit.
  • Environmentally safe, 100% thermoplastic polymer made is recyclable
  • High coefficient of friction, provides non-skid properties
  • Multi-use physical properties
  • Utilized in both safety application and in most Category I Cleanrooms
  • Strong and durable to withstand abrasive floor surfaces
  • Does not contain PVC compounds that can harm environment
  • Thread colors for convenient size identification
  • Serged seams
  • 100 Pairs/Cs.
Specifications
Size X-Large
Material Thermoplastic Polymers
Color White
Sterility Non-sterile
Seam Type Serged
Thread Color Black
Packaging Quantity 100 Pairs/Cs.
Resistance Dry Particulates, Fluid, Light Chemical Splash
Product Type Controlled Environments Shoe Covers