missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ PFL™ N95 Particulate Respirator

Catalog No. 23309788
Change view
Click to view available options
Filter Type:
Particulate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type
23309788 Particulate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23309788 Supplier AlphaProTech Supplier No. MAS 695

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N95 particulate respirator is NIOSH approved and meets WHO/CDC recommended protection levels for many airborne contaminants.

  • NIOSH approved
  • Magic Arch™ holds the mask away from the nose and mouth for comfortable breathing
  • Positive Facial Lock™ (PFL) design helps the wearer create a perfect fit with no gaps that might allow small particles to be inhaled around the edges of the mask
  • Downward vision is not restricted for added safety
  • Teal stripe color

Specifications

Aerosol Type N - Not Resistant to Oil
Certifications/Compliance 21C CFR Part 820
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0457
Color Teal Stripe
Design Type Flat-fold
Disposable Single-use
Filter Type Particulate
Material Composite Fabric
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No
Filter Efficiency 99%
For Use With (Application) Possible exposure to splashing blood, body fluids or other potentially infectious micro sized particles
Latex Free Yes
Model 695
Nosepiece Type M-nose clip
Packaging Type Dispenser Box
Quantity 210/Pk.
Resistant To Fluid 80 mmHg
Sterility Non-sterile
Style Respirator
With Faceseal No
Strap Attachment Type Dual Elastic Straps
FDA Cleared Surgical Mask No
Show More Show Less