missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ AquaTrak™ Shoe Covers

Catalog No. 19811011
Change view
missing translation for 'orderingAttributeHoverText'
:
Black; X-Large
Catalog No.
19811011 Black; X-Large
1 options

Catalog No. 19811011

Supplier: AlphaProTech SH C1103 B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AquaTrak™ material keeps the wearer stable and protected in wet and dry conditions.

  • Anti-skid properties provide amazing slip and fall protection without additional coatings
  • Material is fluid impervious
  • Styles with heat-sealed seams provide complete protection even in standing water
  • Serged seam soles provide great fit and can be used in dry or moist conditions
  • High breathability, low particulate count fabric wears comfortably and completely contains contamination

Specifications

Size X-Large
Material AquaTrak™
Color Black
Closure Type Elastic Top with Ankle Ties
Sole Type Non-skid
Seam Type Serged
Style Butterfly
For Use With (Application) Ideal for superior performance APT and amazing slip and fall protection