Learn More
AlphaProTech Critical Cover™ AlphaAir™ Face Masks
Catalog No. 191304181
Change view
Click to view available options
Color:
Blue
Yellow
Fastening Type:
Earloops
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Fastening Type
191304181 Blue Earloops
19065338 Yellow Earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 191304181 Supplier AlphaProTech Supplier No. BL5005
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Constructed of cool, comfortable hypoallergenic materials that are naturally fluid-resistant. AlphaAir™ face masks feature a 0.1 micron filter with greater than 95% bacterial filtration efficiency.
  • Simple, cool and comfortable design
  • Constructed of materials that are naturally fluid-resistant
  • Highly malleable aluminum nosepiece extends entire length of mask, maximizing comfort and minimizing fogging of eyewear
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Blue
Fastening Type Earloops
Material Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Construction 3-ply
Disposable Single-use
Filter Efficiency ≥98% (PFE), ≥98% (BFE)
Latex Free Yes
Model BL 5005
Packaging Quantity 500/Cs.
Pore Size 0.1 μm
Resistance Fluid
Series AlphaAir™
Show More Show Less