missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Adipose Tissue Section, Human

Numéro de catalogue. S30179
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
S30179 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S30179 Fournisseur Eisco™ Code fournisseur BS18068

Prepared microscope slide of adipose (fatty) tissue. Shows characteristic features of fat vacuoles. Ideal for studying human histology.

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic structures of adipose cells, including fat vacuoles

Spécifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Mounting Type Mounting Medium
Type Microbiology slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Human
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Afficher plus de résultats Afficher moins de résultats