missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acipimox, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 466990050
Specifications
| Acipimox | |
| 5 g | |
| 13-113 | |
| C6 H6 N2 O3 | |
| Powder | |
| ≥98.0% | |
| CC1=CN=CC(C(O)=O)=[N+]1[O-] | |
| 154.13 |
| 51037-30-0 | |
| Soluble in methanol | |
| 180°C to 190°C | |
| 154.12 | |
| White to Almost-white | |
| DNRXJHATQULEHC-UHFFFAOYSA-N | |
| 2-carboxy-6-methylpyrazin-1-ium-1-olate |
Chemical Identifiers
| 51037-30-0 | |
| C6 H6 N2 O3 |
Safety and Handling
Exclamation mark
EINECSNumber : 256-928-3
Recommended Storage : Normal conditions