missing translation for 'onlineSavingsMsg'
Learn More

Acipimox, Thermo Scientific™

Catalog No. p-7238597
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466990010 1 g
AC466990050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466990010 Supplier Thermo Scientific Chemicals Supplier No. 466990010
Only null left

Chemical Identifiers

CAS 51037-30-0
Molecular Formula C6 H6 N2 O3

Specifications

Quantity 1 g
Solubility Soluble in methanol
Merck Index 13-113
Melting Point 180°C to 190°C
Formula Weight 154.12
Physical Form Powder
Color White to Almost-white
Purity ≥98.0%
InChI Key DNRXJHATQULEHC-UHFFFAOYSA-N
SMILES CC1=CN=CC(C(O)=O)=[N+]1[O-]
IUPAC Name 2-carboxy-6-methylpyrazin-1-ium-1-olate
Molecular Weight (g/mol) 154.13
Chemical Name or Material Acipimox
Show More Show Less
Hazard Category Exclamation mark
EINECSNumber 256-928-3
Recommended Storage Normal conditions