CAS RN 51037-30-0
IUPAC Name:
2-carboxy-6-methylpyrazin-1-ium-1-olate
Synonyms:
acipimox
Molecular Weight (g/mol):
154.13
Molecular Formula:
C6H6N2O3
InChi Key:
DNRXJHATQULEHC-UHFFFAOYSA-N
SMILES:
CC1=CN=CC(C(O)=O)=[N+]1[O-]
1
–
2
of
2
results
1
2
5-Methylpyrazine-2-carboxylic Acid 4-Oxide 98.0+%, TCI America™
CAS: 51037-30-0 Molecular Formula: C6H6N2O3 Molecular Weight (g/mol): 154.13 MDL Number: MFCD00865757 InChI Key: DNRXJHATQULEHC-UHFFFAOYSA-N Synonym: acipimox,olbetam,olbemox,5-carboxy-2-methylpyrazine 1-oxide,5-methylpyrazine-2-carboxylic acid 4-oxide,5-methylpyrazinecarboxylic acid 4-oxide,2-carboxy-5-methylpyrazine 4-oxide,5-methyl-2-pyrazinecarboxylic acid 4-oxide,5-methyl-4-oxy-pyrazine-2-carboxylic acid,2-pyrazinecarboxylic acid, 5-methyl-, 4-oxide PubChem CID: 5310993 IUPAC Name: 2-carboxy-6-methylpyrazin-1-ium-1-olate SMILES: CC1=CN=CC(C(O)=O)=[N+]1[O-]