Learn More
Acetate Buffer pH 4.0, For Chlorine, Iodine, Certified, LabChem™
Supplier:  LabChem LC100305
 View more versions of this product
Catalog No. LC100305
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Acetate Buffer pH 4.0
64-19-7 , 7732-18-5 , 6131-90-4
45.5,32.5,22
4.0 ±0.1
20 L
CH3COOHNa·3H2O
Traceable to NIST
136.08
For Chlorine, Iodine
Colorless
Carbon monoxide, Carbon dioxide
Liquid
Passes Test
Cubitainer™
Soluble in water
Certified
Product Suggestions
Product Suggestions