missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer pH 4.0, For Chlorine, Iodine, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Acetate Buffer pH 4.0 |
| Name Note | For Chlorine, Iodine |
| CAS | 64-19-7, 7732-18-5, 6131-90-4 |
| Color | Colorless |
| CAS Max % | 45.5, 32.5, 22 |
| Decomposition Information | Carbon monoxide, Carbon dioxide |
| pH | 4.0 ±0.1 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Identification | Passes Test |
| Show More |