missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ AR425 pH/Ion Meter pH Kit
Description
- Large 7-in., high-resolution color display with capacitive touchscreen operable with gloves.
- One to five point pH and Ion calibration with automatic buffer/standard recognition.
- Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
- Stores 10 calibration logs per parameter with date and time.
- Stores 5000 data log sets with date and time.
- Outputs to a printer, computer, or USB flash drive for long-term data retention.
- Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
- Stirs samples with meter-controlled and meter-powered stirrer probe.
Spécifications
Spécifications
| Accuracy (mV) | ±0.2 mV or ±0.05% of the reading, whichever is greater |
| Accuracy (pH) | ±0.002 pH |
| Backlight Option | Yes |
| Calibration | pH |
| Calibration Points | 1 |
| Certifications/Compliance | CE, TUV 3-1, FCC Class A |
| Data Points | 5000 with Date and Time Stamp |
| Channels | 1 |
| Display Type | 7-in. High-resolution Color Graphic LCD |
| IP Rating | IP54 |
| Afficher plus de résultats |