missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Non Binding Microplates

Description
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
- Significantly increase assay sensitivity
Spécifications
Spécifications
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 25 to 340 μL |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |
| Afficher plus de résultats |