Learn More
Greiner Bio-One 96-well Half Area ELISA Microplates
GREENER_CHOICE
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
Supplier:  Greiner Bio-One 675161
 Afficher plus de versions de ce produit
Numéro de catalogue. 07000082
Il en reste null
Description
Description
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Crystal clear polystyrene
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Well-to-well spacing: 9mm
  • Specifically tre
    Certifications
    DIN ISO 9001
Spécifications
Identifiants chimiques
Half-Area
Clear
96
8 x 12
Polystyrene
40/Cs.
Spécifications
No
Without Lid
Clear
96
Non-treated
Round, Flat Bottom (F-well)
10
Clear
Half-Area
8 x 12
Polystyrene
Sterile
199 μL
40
40/Cs.
Chimney
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS