missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Half Area ELISA Microplates

For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
$687.39 - $743.71
Specifications
| Format | Half-Area |
|---|---|
| Array | 8 x 12 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Catalog Number | Mfr. No. | Sterility | Surface Treatment | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Surface Treatment | Price | Quantity & Availability | ||||
07000091
![]() |
Greiner Bio-One
675101 |
Non-sterile | Non-treated |
Case of 40 Each for $690.54
|
|
||||
07000720
![]() |
Greiner Bio-One
675061 |
Non-sterile | Microlon 600 High Binding |
Case of 40 Each for $743.71
|
|
||||
07000082
![]() |
Greiner Bio-One
675161 |
Sterile | Non-treated |
Case of 40 Each for $690.04
|
|
||||
07000084
![]() |
Greiner Bio-One
675001 |
Non-sterile | Microlon Medium Binding |
Case of 40 Each for $687.39
|
|
||||
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Specifically tre
Certifications
DIN ISO 9001
Specifications
| Half-Area | |
| Clear | |
| 96 |
| 8 x 12 | |
| Polystyrene | |
| 40/Cs. |