Learn More
Cole-Parmer™ Adapters and Clamps for High-Energy Mixer/Mill™ Grinders
Use adapters to run multiple samples or to hold vial securely in place during grinding.
Supplier:  Cole-Parmer™ SPX8010
 Afficher plus de versions de ce produit
Numéro de catalogue. 8010
Il en reste null
Description
Description
  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ Cryogenic Grinders
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinder with a wide range of adapters and clamps.
Spécifications
Identifiants chimiques
Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples
8000 D and 8000 M Mixer or Mill
Spécifications
Multiple Sample Adapter
Ideal for High Throughput Applications That Involve Processing Multiple Smaller Samples
8000 D and 8000 M Mixer or Mill
7 x 0.5 in. dia. Vials
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS