missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline Solution, LabChem™

Numéro de catalogue. LC155801
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
LC155801 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. LC155801 Fournisseur LabChem Code fournisseur LC155801
Il en reste null

Identifiants chimiques

CAS 7732-18-5 , 64-19-7 , 148-24-3
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Spécifications

Color Yellow
CAS Max % 91.76, 6.24, 2
Linear Formula HOC6H3N:CHCH:CH
Density 1.01g/mL
Solubility Information Soluble in water
Formula Weight 145.16
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.01g/mL
Viscosity 1.1cSt
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material 8-Hydroxyquinoline Solution
Afficher plus de résultats Afficher moins de résultats
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature