accessibility menu, dialog, popup

UserName

OHN

CAS RN 148-24-3

IUPAC Name: quinolin-8-ol
Synonyms: 8 hydroxyquinoline oxyquinoline (product) bioquin caswell no. 719 oxyquinol leioderm riemser brand of oxyquinoline sulfate phenopyridine fennosan 8-oxyquinol
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: MCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=CC=C1
Quantity 
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (9)
  • (5)
Physical Form 
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
Grade 
  • (5)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (16)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (11)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (9)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)