missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 8 cal Coverall Arc Flash Kit (09)

Catalog No. 17001829
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001829 5X-Large
17001794 Small
17001799 Medium
17001804 Large
17001809 X-Large
17001814 2X-Large
17001819 3X-Large
17001824 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001829 Supplier National Safety Apparel™ Supplier No. KIT2CV08B5X09

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The premium choice for superior comfort, improved safety and above average durability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper

Specifications

Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 8 cal/cm²
Ankle Style Open
Size 5X-Large
Color Navy
Material Cotton, Nylon
Suspender None
Closure Type Zipper
Seam Type Stitched
Show More Show Less