Learn More
Eisco™ 6 Pack of Graduated Beakers with No Spout
Berzelius Beakers: Borosilicate glass, tall form, ideal for titrations, white enamel markings, DIN/ISO standards, no pouring spout, microwavable, autoclavable, high chemical durability, low temperature gradient, withstands high temperatures and shocks.
Supplier:  Eisco™ CH2006PK6
 Afficher plus de versions de ce produit
Numéro de catalogue. S80041
Il en reste null
Description
Description
  • TALL FORM || Uniform walls of Berzelius tall-form beakers are ideal for heating and stirring during titrations. Their smaller diameter makes them easier to handle than low form beakers of the same volume
  • NO SPOUT || Tall form beakers without spout are ideal for containing and storing samples
  • GRADUATIONS || 500 mL capacity beakers feature white enamel markings with 50 mL graduations labeled every 100 mL. Conforms to DIN 12231 and ISO 3819 standards
  • INDUSTRIAL QUALITY || Made from heavy-duty borosilicate 3.3 glass. Microwavable and autoclavable.
Spécifications
Identifiants chimiques
Borosilicate Glass (3.3)
Autoclavable
Yes
Berzelius
Non-sterile
Reusable
White Enamel Scale
Spécifications
400 mL
Non-sterile
Reusable
White Enamel Scale
Borosilicate Glass (3.3)
Autoclavable
Yes
Berzelius
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS